Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin 17 Mouse anti-Human, Unconjugated, Clone: 7C5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT152625UG
Description
Cadherin 17 Monoclonal antibody specifically detects Cadherin 17 in Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications
Cadherin 17 | |
Monoclonal | |
Unconjugated | |
Intestinal peptide-associated transporter HPT-1;Liver-intestine cadherin;LI-cadherin | |
Mouse | |
92.22 kDa calculated. | |
25 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
7C5 | |
NP_001138135 | |
CDH17 | |
Recombinant human Cadherin 17-Fc Protein. | |
Protein G Purified | |
Cell Structure | |
Clone 7C5 is a a mouse monoclonal antibody that detects human Cadherin 17. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction