Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin 17 Mouse anti-Human, Unconjugated, Clone: 7C5, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $457.74
Specifications
Antigen | Cadherin 17 |
---|---|
Clone | 7C5 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT15262MI
|
MilliporeSigma
MABT152625UG |
25 μg |
Each for $182.31
|
|
|||||
MABT15261MI
|
MilliporeSigma
MABT1526100UG |
100 μg |
Each for $457.74
|
|
|||||
Description
Cadherin 17 Monoclonal antibody specifically detects Cadherin 17 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Cadherin 17 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Intestinal peptide-associated transporter HPT-1;Liver-intestine cadherin;LI-cadherin | |
Recombinant human Cadherin 17-Fc Protein. | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
92.22 kDa calculated. |
7C5 | |
Monoclonal | |
Purified | |
Cell Structure | |
NP_001138135 | |
CDH17 | |
IgG2a κ | |
Protein G Purified | |
Clone 7C5 is a a mouse monoclonal antibody that detects human Cadherin 17. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title