Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-23 Antibody, Novus Biologicals™
SDP

Catalog No. NBP120898 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120898 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120898 Supplier Novus Biologicals Supplier No. NBP120898
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Cadherin-23 Polyclonal specifically detects Cadherin-23 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Cadherin-23
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q9H251
Gene Alias cadherin related 23, cadherin-23, cadherin-like 23, cadherin-related 23, cadherin-related family member 23, CDHR23, DKFZp434P2350, FLJ00233, FLJ36499, KIAA1774, KIAA1812, MGC102761, Otocadherin, USH1D
Gene Symbols CDH23
Host Species Goat
Immunogen Peptide with sequence C-YNISLYENVTVGTS corresponding to internal region according to NP_071407.3, NP_443068.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64072
Test Specificity This antibody is expected to recognize both reported isoforms (NP_071407.3; NP_443068.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.