Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-6/KCAD Antibody (09), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB388724 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB388724 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB388724 Supplier Novus Biologicals Supplier No. NBP312779AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherin-6/KCAD Monoclonal antibody specifically detects Cadherin-6/KCAD in Human samples. It is validated for ELISA

Specifications

Antigen Cadherin-6/KCAD
Applications ELISA
Classification Monoclonal
Clone 09
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias cadherin 6, type 2, K-cadherin (fetal kidney), cadherin, fetal kidney, cadherin-6, KCAD, K-cadherin, K-cadherin (fetal kidney)
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cadherin-6/KCAD (Accession#: NP_004923.1; Met1-Ala615).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1004
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.