Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadherin-6/KCAD Antibody (2B6), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP197689R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197689R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197689R Supplier Novus Biologicals Supplier No. NBP197689R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cadherin-6/KCAD Monoclonal specifically detects Cadherin-6/KCAD in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Cadherin-6/KCAD
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2B6
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cadherin 6, type 2, K-cadherin (fetal kidney), cadherin, fetal kidney, cadherin-6, KCAD, K-cadherin, K-cadherin (fetal kidney)
Gene Symbols CDH6
Host Species Mouse
Immunogen 2B6 is a mouse monoclonal IgG1 antibody obtained by fusion of SP2/0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with affinity purified extracellular domain of Human cadherin-6-GST fusion protein.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1004
Test Specificity 2B6 recognizes the extracellular domain of cadherin-6.
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.