Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-6/KCAD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32122025UL
Description
Cadherin-6/KCAD Polyclonal antibody specifically detects Cadherin-6/KCAD in Human samples. It is validated for ImmunofluorescenceSpecifications
| Cadherin-6/KCAD | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| cadherin 6, type 2, K-cadherin (fetal kidney), cadherin, fetal kidney, cadherin-6, KCAD, K-cadherin, K-cadherin (fetal kidney) | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ADVGENAEIEYSITDGEGLDMFDVITDQETQEGIITVKKLLDFEKKKVYTLKVEASNPYVEPRFLYLGPFKDSATVRIVVEDVDEPPVFSKLAYILQIREDAQINTTIGSVTAQDPDAARNP | |
| 25 ul | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 1004 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction