Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cadherin-6/KCAD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$322.50 - $547.00
Specifications
Antigen | Cadherin-6/KCAD |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cadherin-6/KCAD Polyclonal antibody specifically detects Cadherin-6/KCAD in Human samples. It is validated for ImmunofluorescenceSpecifications
Cadherin-6/KCAD | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
1004 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
cadherin 6, type 2, K-cadherin (fetal kidney), cadherin, fetal kidney, cadherin-6, KCAD, K-cadherin, K-cadherin (fetal kidney) | |
This antibody was developed against Recombinant Protein corresponding to amino acids: ADVGENAEIEYSITDGEGLDMFDVITDQETQEGIITVKKLLDFEKKKVYTLKVEASNPYVEPRFLYLGPFKDSATVRIVVEDVDEPPVFSKLAYILQIREDAQINTTIGSVTAQDPDAARNP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title