Learn More
Epredia™ Lab Vision™ Cadherin E Ab-4, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cadherin E Ab-4, Mouse Monoclonal Antibody.
$947.00
Specifications
Antigen | Cadherin E/E-Cadherin Ab-4 |
---|---|
Clone | NCH-38 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS1862S1
|
Epredia™
MS1862S1 |
Concentrated | 500 μL |
Each for $947.00
|
|
||||
MS1862S0
|
Epredia™
MS1862S0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS1862R1
|
Epredia™
MS1862R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
MS1862R7
|
Epredia™
MS1862R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
E-cadherin (uvomorulin, cell-CAM120/80) is a calcium-dependent cell adhesion molecule expressed predominately in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.Host Species: Mouse
Clone: NCH-38
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: E-cadherin and GST recombinant protein
Molecular Weight: 120 kDa
Positive Control: Breast CA
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Note:
USA: IVD; Int'l: IVD
Specifications
Cadherin E/E-Cadherin Ab-4 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
IgG2a κ |
NCH-38 | |
Monoclonal | |
Mouse | |
Junction Protein | |
E-cadherin and GST recombinant protein | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.