Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ E-Cadherin Rabbit Monoclonal Antibody

Ensure accurate and reliable results in immunohistochemistry applications with Epredia™ Cadherin E, Rabbit Monoclonal Antibody.

$327.00 - $2159.00

Specifications

Antigen E-Cadherin
Clone EP700Y
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RM2100S0
View Documents
Epredia™
RM2100S0
Concentrated 100 μL
Each for $327.00
Only null left
Add to Cart
 
RM2100S
View Documents
Epredia™
RM2100S
Concentrated 1 mL
Each for $2,159.00
Only null left
Add to Cart
 
RM2100R7
View Documents
Epredia™
RM2100R7
Prediluted 7 mL
Each for $1,423.00
Only null left
Add to Cart
 
RM2100RQ
View Documents
Epredia™
RM2100RQ
Ready-to-Use 12 mL
Each for $1,797.00
Only null left
Add to Cart
 
RM2100R1
View Documents
Epredia™
RM2100R1
Prediluted 1 mL N/A N/A N/A
Description

Description

E-cadherin (uvomorulin, cell-CAM120/80) is a calcium-dependent cell adhesion molecule expressed predominantly in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis. E-cadherin is used in distinguishing LCIS from DCIS and that of ILC from IDC in indeterminate cases.

Host Species: Rabbit

Clone: EP700Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from the fifth cadherin domain of human cadherin

Molecular Weight: 135/80kDa

Positive Control: Breast Carcinoma

Cellular Localization: Cell Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

E-Cadherin
Immunohistochemistry (Paraffin)
Unconjugated
IVD
A synthetic peptide derived from the fifth cadherin domain of human cadherin
EP700Y
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.