Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CADPS Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB356011 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356011 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356011 Supplier Novus Biologicals Supplier No. NBP177324PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CADPS Polyclonal antibody specifically detects CADPS in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CADPS
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Ca++-dependent secretion activator, CADPS1, Calcium-dependent activator protein for secretion 1, calcium-dependent secretion activator 1, CAPS-1, CAPS1Ca2+-dependent secretion activator, CAPSCa2+-dependent activator protein for secretion, KIAA1121Ca2+-regulated cytoskeletal protein
Host Species Rabbit
Immunogen Antibody was raised against a 21 amino acid synthetic peptide near the amino terminus of the human CAPS1. The immunogen is located within amino acids 90 - 140 of CAPS1. Amino Acid Squence: SEKEKEELERLQKEEEERKKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8618
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.