Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calbindin D-28K Antibody (CALB1/2364), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB381709 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381709 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381709 Supplier Novus Biologicals Supplier No. NBP308309JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human samples. It is validated for ELISA, Protein Array

Specifications

Antigen Calbindin D-28K
Applications ELISA, Protein Array
Classification Monoclonal
Clone CALB1/2364
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type
Host Species Mouse
Immunogen Recombinant fragment (around aa 7-96) of human Calbindin D-28K protein (exact sequence is proprietary) (Uniprot: P05937)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 793
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.