Learn More
Calbindin D-28K Mouse anti-Human, Clone: CALB1/2782, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Calbindin D-28K |
| Applications | ELISA, Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | CALB1/2782 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.