Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Y-27632

Catalog No. p-7060183 Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
100 mg

Catalog No. 68-800-01MG

Supplier: MilliporeSigma™ 6880001MG

Only null left
Add to Cart
Add to Cart

Y-27632A, CAS 146986-50-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Purity 95% by HPLC
Solubility H2O
Molecular Formula C14H21N3O·2HCl·H2O
CAS 146986-50-7
Content And Storage Hygroscopic
Protect from Light
Protein p160 Rho-associated protein kinases (ROCK)
Quantity 1 mg
Molecular Weight (g/mol) 338.3g/mol
Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.