Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Nucleozin
Shop All MilliporeSigma Products
Click to view available options
Quantity:
25 mg
Description
EMD Millipore Calbiochem™ Nucleozin reduces fatality after H5N1 infection in mice in vivo by preventing nucleoprotein (NP) nuclear accumulation.
- Cell-permeable isoxazolylpiperazine compound
- Replication in MDCK cells in vitro (EC50 = 69, 160, 330nM)
- Significantly reduce fatality by NP aggregation induction during early stages of viral infection
- Assay: ≥95% by HPLC
- Molecular Formula: C21H19ClN4O4
- Packaging: Glass Bottle
Recommended Storage
2° to 8°C
Specifications
Specifications
| Description | Nucleozin |
| Quantity | 25 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction