Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Y-27632
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
100 mg
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Specifications
| Color | White |
| Purity | 95% by HPLC |
| Solubility | H2O |
| Molecular Formula | C14H21N3O·2HCl·H2O |
| CAS | 146986-50-7 |
| Content And Storage | Hygroscopic Protect from Light |
| Protein | p160 Rho-associated protein kinases (ROCK) |
| Quantity | 100 mg |
| Molecular Weight (g/mol) | 338.3g/mol |
| Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction