Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcineurin A Antibody (CALNA/2353), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB380837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380837 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380837 Supplier Novus Biologicals Supplier No. NBP308395JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calcineurin A Monoclonal antibody specifically detects Calcineurin A in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Calcineurin A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CALNA/2353
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1CALNA1, EC 3.1.3.16, PPP2BCCN1, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Host Species Mouse
Immunogen Recombinant full-length human Calcineurin A protein (Uniprot: Q08209)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Protein Phosphatase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5530
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.