Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcineurin A Rabbit anti-Human, Mouse, Rat, Clone: 5I6H8, Novus Biologicals™
SDP

Catalog No. NB167854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167854 20 μg
NB167853 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167854 Supplier Novus Biologicals Supplier No. NBP31638120UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Calcineurin A Monoclonal antibody specifically detects Calcineurin A in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Calcineurin A
Applications Western Blot
Classification Monoclonal
Clone 5I6H8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1CALNA1, EC 3.1.3.16, PPP2BCCN1, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 422-521 of human Calcineurin A (Q08209).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Protein Phosphatase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5530
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.