Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcineurin B Antibody (BE2.1), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB392270 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392270 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392270 Supplier Novus Biologicals Supplier No. NBP308451UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calcineurin B Monoclonal antibody specifically detects Calcineurin B in Human, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Calcineurin B
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BE2.1
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias calcineurin subunit B type 1, CNA2, CNB1, Protein phosphatase 2B regulatory subunit 1, protein phosphatase 2B regulatory subunit B alpha, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform, Protein phosphatase 3 regulatory subunit B alpha isoform 1, protein phosphatase 3, regulatory subunit B, alpha, type I (19kDa)
Host Species Mouse
Immunogen Recombinant full-length human Calcineurin B protein (Uniprot: P63098)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5534
Target Species Human, Xenopus species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.