Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcineurin B Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB060713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB060713 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB060713 Supplier Novus Biologicals Supplier No. NBP299441DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calcineurin B Polyclonal specifically detects Calcineurin B in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Calcineurin B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias calcineurin subunit B type 1, CNA2, CNB1, Protein phosphatase 2B regulatory subunit 1, protein phosphatase 2B regulatory subunit B alpha, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform, Protein phosphatase 3 regulatory subunit B alpha isoform 1, protein phosphatase 3, regulatory subunit B, alpha, type I (19kDa)
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse Calcineurin B.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5534
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.