Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcitonin R Antibody (462802), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB369709 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369709 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369709 Supplier Novus Biologicals Supplier No. FAB4614W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calcitonin R Monoclonal specifically detects Calcitonin R in Human samples. It is validated for CyTOF, Flow Cytometry, Gene Knock-Out.

Specifications

Antigen Calcitonin R
Applications CyTOF, Flow Cytometry, Gene Knock-Out
Classification Monoclonal
Clone 462802
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias calcitonin receptor, CRT, CTR, CT-R, CTR1
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Calcitonin R, Ala25-Ala474, Accession # NP_001733
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 799
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.