Learn More
Calcium carbonate, Puratronic™, 99.99% (metals basis)
Description
Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It forms the major component of ceramic tile adhesives. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 800°C (decomposition) |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Merck Index | 14,1657 |
| Solubility Information | Very low solubility in water. |
| Formula Weight | 100.09 |
| Percent Purity | 99.99% |
| Grade | Puratronic™ |
| Odor | Odorless |
| Show More |
Safety and Handling
| EINECSNumber | 207-439-9 |
| RTECSNumber | FF9335000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.