Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Chloride, Dihydrate, BiotechGrade, 99-103%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Calcium Chloride, Dihydrate, BiotechGrade is a white deliquescent compound which is commonly used as a reagent in biochemistry. Biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability rely on calcium ions. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.
Specifications
Specifications
pH | 4.5- 9.2 |
CAS Max % | 100% |
Quantity | 500 g |
Percent Purity | 99 to 103% |
Grade | Biotech |
Packaging | Poly Bottle |
Appearance | White crystals |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction