Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride Dihydrate, puriss., Honeywell Fluka™

Catalog No. 6001916 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
2.5 kg
15 kg
6 x 2.5 kg

Puriss., meets analytical specification of FCC, Ph. Eur., USP, E509, 99-103%, ≤0.0001% Al

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2 · 2H2O
MDL Number MFCD00149613

Specifications

Quantity 15 kg
UN Number NONH for all modes of transport
Formula Weight 147.01g/mol
Percent Purity 99 to 103%
Grade puriss.
Chemical Name or Material Calcium Chloride Dihydrate, Meets analytical specification of Ph. Eur.USP, FCC
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 233-140-8
RTECSNumber EV9810000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.