Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Dihydrate, Reagent, ACS, 99-105%, Spectrum™ Chemical
SDP

Catalog No. 18601268 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

C1096, 10035-04-8, CaCl2·2H2O

Spectrum™ Chemical Calcium Chloride, Dihydrate, Reagent, ACS is a analytical grade chemical that is packaged in laboratory size bottles to production drum quantities. As an ACS grade Reagent, Spectrum Chemical manufactured Calcium Chloride is used as the quality standard against which other Calcium Chloride substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2H4O2
Molecular Weight (g/mol) 147.01
InChI Key LLSDKQJKOVVTOJ-UHFFFAOYSA-L
IUPAC Name calcium dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ca++]

Specifications

pH 4.5- 8.5
CAS Max % 100%
Quantity 2.5 kg
Percent Purity 99 to 105%
Grade ACS
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.