Learn More
Calcium D-gluconate gel, 2.5% w/w aq. soln.
Description
Calcium D-gluconate gel is used to treat hydrofluoric acid burns. It is used as a cardioprotective agent in hyperkalemia, antidote for black widow spider envenomation and an antidote for magnesium sulfate toxicity. It is also involved in the treatment of hypocalcemia. Further, it is used as skin conditioning as well as provides the source of calcium ions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 30 g |
| Solubility Information | Slowly soluble in water. Insoluble in alcohol,acetic acid and organic solvents. |
| Formula Weight | 430.37 |
| Concentration or Composition (by Analyte or Components) | 2.5% w/w aq. soln. |
| Physical Form | Gel |
| Chemical Name or Material | Calcium D-gluconate gel |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.