Learn More
Calcium fluoride, 97%
Description
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.
Solubility
Soluble in ammonium salts. Soluble in water (slightly).
Notes
Store at room temperature. Separated from mineral acids. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
 
            Specifications
Specifications
| Boiling Point | ∼2500°C | 
| Melting Point | 1403°C | 
| Quantity | 250 g | 
| Merck Index | 14,1667 | 
| Solubility Information | Soluble in ammonium salts. Soluble in water (slightly). | 
| Formula Weight | 78.08 | 
| Percent Purity | 97% | 
| Odor | Odorless | 
| Refractive Index | 1.434 | 
| Density | 3.18 | 
| Show More | 
Safety and Handling
| Hazard Category | Warning | 
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. | 
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. | 
| EINECSNumber | 232-188-7 | 
| RTECSNumber | EW1760000 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
 
                                