Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium granules, -9 mesh, 98.8% (metals basis), Thermo Scientific Chemicals

Catalog No. aa4165318
Click to view available options
Quantity:
50 g
250 g
1 kg

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Color Silver-White
Appearance Silvery-white granules
Density 1.54g/cm3
Quantity 50 g
Mesh Size -9 Mesh
Odor Odorless
Vapor Pressure 0 hPa
Form Granular
Assay Percent Range 98.8% (Metals basis)
Chemical Name or Material Calcium
Hazard Category H261
Precautionary Statement P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 213-179-5
RTECSNumber EV8040000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.