Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calcium homeostasis modulator 2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB320165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320165 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320165 Supplier Novus Biologicals Supplier No. NBP298589JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

calcium homeostasis modulator 2 Polyclonal antibody specifically detects calcium homeostasis modulator 2 in Human samples. It is validated for Western Blot

Specifications

Antigen calcium homeostasis modulator 2
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias calcium homeostasis modulator 2, calcium homeostasis modulator protein 2, FAM26Bfamily with sequence similarity 26, member B, Protein FAM26B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human calcium homeostasis modulator 2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 51063
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.