Learn More
Calcium hypochlorite, tech.
Description
Deodorant, oxidizing agent, bleaching agentCalcium hypochlorite is used for swimming pool sanitation and other water treatment plants. It is also used to disinfect drinking water, equipments in the industry and surfaces and equipments in kitchens. Further, it is used to bleach textiles and paper. It is also employed in bathroom cleaners and laundry detergents. It is used as an oxidizing agent in organic chemistry. It is involved in the cleavage of glycols, alfa-hydroxy carboxylic acids and keto acids to yield fragmented aldehydes or carboxylic acids respectively. It plays an important role in the haloform reaction to prepare chloroform.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100°C |
| Color | White |
| Physical Form | Granules |
| Quantity | 250 g |
| Solubility Information | Soluble in water. |
| SMILES | [O-]Cl.[O-]Cl.[Ca+2] |
| IUPAC Name | calcium;dihypochlorite |
| Molecular Weight (g/mol) | 142.976 |
| Formula Weight | 142.98 |
| Appearance | White granular powder |
| Show More |
Safety and Handling
| Hazard Category | H272-H290-H302-H314-H335 |
| Precautionary Statement | P210-P220-P221-P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378-P390-P501c |
| DOTInformation | DOT Class: 5.1, Packaging Group: II |
| EINECSNumber | 231-908-7 |
| RTECSNumber | NH3485000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.