Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium phosphate, monobasic, 90%, pure

Catalog No. AC378610010
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC378610010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC378610010 Supplier Thermo Scientific Chemicals Supplier No. 378610010
Only null left
Add to Cart

CAS: 7758-23-8 | CaH4O8P2 | 234.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-23-8
Molecular Formula CaH4O8P2
Molecular Weight (g/mol) 234.05
MDL Number MFCD00010898
InChI Key YYRMJZQKEFZXMX-UHFFFAOYSA-L
Synonym calcium biphosphate, monobasic calcium phosphate, calcium dihydrogen phosphate, acid calcium phosphate, primary calcium phosphate, monocalcium orthophosphate, c 38 phosphate, calcium diorthophosphate, monocalcium phosphate, monobasic, calcium dihydrogen orthophosphate
PubChem CID 24454
ChEBI CHEBI:35433
IUPAC Name calcium;dihydrogen phosphate
SMILES OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2]

Specifications

Quantity 1 kg
Assay Percent Range 88% min. (Complexometry)
Linear Formula Ca(H2PO4)2
Merck Index 15,1695
Solubility Information Solubility in water: insoluble
Formula Weight 234.05
Percent Purity 90%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Calcium phosphate, monobasic
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
EINECSNumber 231-837-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.