Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Phosphate Monobasic, Monohydrate
SCI_ED

Catalog No. S25228 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Packaging:
Solid

Catalog No. S25228

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 10031-30-8
Molecular Formula CaH6O9P2
Molecular Weight (g/mol) 252.065
InChI Key ZBZJARSYCHAEND-UHFFFAOYSA-L
Synonym calcium phosphate, monobasic, monohydrate, calcium dihydrogenphosphate hydrate, calcium phosphate monobasic monohydrate, primary calcium phosphate monohydrate, calcium dihydrogenphosphate monohydrate, calcium dihydrogen phosphate monohydrate, calcium bis dihydrogenphosphate monohydrate, monobasic calcium phosphate hydrate jp17, calcium bis dihydrogen phosphate monohydrate, calcium hydrate didihydrogen phosphate
PubChem CID 10198893
IUPAC Name calcium;dihydrogen phosphate;hydrate
SMILES O.OP(=O)(O)[O-].OP(=O)(O)[O-].[Ca+2]

Specifications

Quantity 100 g
Assay Percent Range >85%
Linear Formula Ca(H2PO4)2·H2O
Formula Weight 252.07
Packaging Solid
Density 2.22g/mL
Chemical Name or Material Calcium Phosphate Monobasic, Monohydrate
ShelfLife 2 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.