Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium-sensing R/CaSR Antibody (HL 1499), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB1001830Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001830Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001830Z Supplier Novus Biologicals Supplier No. NB1001830AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calcium-sensing R/CaSR Monoclonal specifically detects Calcium-sensing R/CaSR in Human, Mouse, Rabbit, Rat samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot.

Specifications

Antigen Calcium-sensing R/CaSR
Applications ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone HL 1499
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Gene Accession No. P41180
Gene Alias calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441
Gene Symbols CASR
Host Species Mouse
Immunogen N-terminal region of human Calcium Sensing Receptor. [UniProt# P41180]
Molecular Weight of Antigen 130 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Extracellular Matrix, GPCR, Growth and Development, Lipid and Metabolism, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 846
Target Species Human, Mouse, Rat, Rabbit, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.