Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium-sensing R/CaSR Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB343538 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343538 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343538 Supplier Novus Biologicals Supplier No. NBP241271MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calcium-sensing R/CaSR Polyclonal antibody specifically detects Calcium-sensing R/CaSR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Calcium-sensing R/CaSR
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias calcium-sensing receptor, CAR, CaSR, EIG8, extracellular calcium-sensing receptor, FHH, FIH, GPRC2AHHC, HHC1, hypocalciuric hypercalcemia 1, NSHPT, parathyroid Ca(2+)-sensing receptor 1, Parathyroid cell calcium-sensing receptor, PCaR1, PCAR1MGC138441
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid peptide near the amino terminus of human CASR. The immunogen is located within the first 50 amino acids of CASR. Amino Acid Squence: TSAYGPDQRAQKKGD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix, GPCR, Growth and Development, Lipid and Metabolism, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 846
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.