Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caldesmon/CALD1 Antibody (CALD1/820), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP247818A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP247818A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP247818A30 Supplier Novus Biologicals Supplier No. NBP247818AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caldesmon/CALD1 Monoclonal specifically detects Caldesmon/CALD1 in Human, Rat (Negative) samples. It is validated for Western Blot.

Specifications

Antigen Caldesmon/CALD1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone CALD1/820
Conjugate Alexa Fluor 350
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22
Gene Symbols CALD1
Host Species Mouse
Immunogen Recombinant full-length human Caldesmon/CALD1 protein (Uniprot: Q05682)
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 800
Test Specificity Recognizes a protein of 150kDa, which is identified as the high molecular weight variant of Caldesmon. Two closely related variants of human caldesmon have been identified which are different in their electrophoretic mobility and cellular distribution. The h-caldesmon variant (12080kDa) is found in non- muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle. This monoclonal antibody recognizes only the 150kDa variant (h-caldesmon) in Western blots of human aortic media extracts and is unreactive with fibroblast extracts from cultivated human foreskin. Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction.
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.