Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Calhex 231 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
Negative allosteric modulator of the calcium sensing receptor (CaSR). Blocks increases in [3H]inositol phosphate levels elicited by wild-type hCasR activation (IC50= 0.39 μM in transiently transfected HEK293 cells).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 443.41 |
| Purity | >98% |
| Molecular Weight (g/mol) | 406.954 |
| Chemical Name or Material | Calhex 231 hydrochloride |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction