Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CALHM1 Mouse anti-Human,Mouse, Unconjugated, Clone: 32C2, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | CALHM1 |
---|---|
Clone | 32C2 |
Applications | Dot Blot, Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN25212MI
|
MilliporeSigma
MABN252125UL |
25 μL |
Each for $189.29
|
|
|||||
MABN25211MI
|
MilliporeSigma
MABN2521100UL |
100 μL |
Each for $477.80
|
|
|||||
Description
CALHM1 Monoclonal antibody specifically detects CALHM1 in Human,Mouse samples. It is validated for Dot Blot, Immunohistochemistry, Western Blotting.Specifications
CALHM1 | |
Dot Blot, Immunohistochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Calcium homeostasis modulator protein 1;Protein FAM26C | |
Recombinant fragment corresponding to 144 amino acids from the C-terminal region of human Calcium homeostasis modulator protein 1 (CALHM1). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼46 kDa observed; 38.26 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
32C2 | |
Monoclonal | |
Purified | |
Neuroscience | |
NP_001001412.3 | |
CALHM1, FAM26C | |
IgG2a κ | |
Protein G Purified | |
Clone 32C2 is a mouse monoclonal antibody that detects Calcium homeostasis modulator protein 1 (CALHM1). It targets an epitope within 11 amino acids from the C-terminal region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title