Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calicivirus Antibody (FCV1-43), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB10063286C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063286C 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063286C Supplier Novus Biologicals Supplier No. NB10063286C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calicivirus Monoclonal specifically detects Calicivirus in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Calicivirus
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone FCV1-43
Conjugate DyLight 650
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FCV
Host Species Mouse
Immunogen Calcivirus viral capsid protein (63 kDa)
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-63286 specifically recognizes feline calicivirus, and does not cross-react with FIV or FeLV.The antibody detects a 63kD band in Western blots, representing the viral capsid protein. It may also be used in immunofluorescence techniques to detect viral infection, and has neutralising activity against feline calicivirus.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.