Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calmodulin 2 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB322877 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322877 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322877 Supplier Novus Biologicals Supplier No. NBP299992PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calmodulin 2 Polyclonal antibody specifically detects Calmodulin 2 in Human samples. It is validated for ELISA

Specifications

Antigen Calmodulin 2
Applications ELISA
Classification Polyclonal
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias CALM, CALM1, CALM3, CALML2, calmodulin, calmodulin 2 (phosphorylase kinase, delta), CAM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMII, CAMIII, FLJ99410, LP7057 protein, PHKD, PHKD2, phosphorylase kinase delta
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Calmodulin 2 (Accession#: P0DP24; Met1-Lys149)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 805
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.