Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calmodulin Antibody (2D1), PE, Novus Biologicals™
SDP

Catalog No. NB1202860PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1202860PE 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1202860PE Supplier Novus Biologicals Supplier No. NB1202860PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calmodulin Monoclonal specifically detects Calmodulin in Human, Mouse, Rat, Bacteria, Bovine, Chicken samples. It is validated for Flow Cytometry.

Specifications

Antigen Calmodulin
Applications Flow Cytometry
Classification Monoclonal
Clone 2D1
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CALM, CALM2, CALM3, CALML2, calmodulin, calmodulin 1 (phosphorylase kinase, delta), CaM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMI, CAMIII, DD132, PHKDCAM, phosphorylase kinase, delta subunit
Gene Symbols CALM1
Host Species Mouse
Immunogen Calmodulin purified from Dictyostelium discoideum.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 801
Test Specificity This detects calmodulin. It does not detect parvalbumin, tropinin, S-100, or myosin light chain kinase (MLCK).
Target Species Human, Mouse, Rat, Bacteria, Bovine, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.