Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calmodulin Antibody (C16T), Novus Biologicals™
SDP

Catalog No. NB019465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019465 0.025 mL
NBP229791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019465 Supplier Novus Biologicals Supplier No. NBP2297910.025ML
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Calmodulin Monoclonal specifically detects Calmodulin in Vertebrate samples. It is validated for Western Blot.

Specifications

Antigen Calmodulin
Applications Western Blot
Classification Monoclonal
Clone C16T
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation 70mM Tris-Glycine 105mM NaCl and 30% Glycerol with Sodium Azide
Gene Alias CALM, CALM2, CALM3, CALML2, calmodulin, calmodulin 1 (phosphorylase kinase, delta), CaM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMI, CAMIII, DD132, PHKDCAM, phosphorylase kinase, delta subunit
Gene Symbols CALM1
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to the Calmodulin Binding Protein (CBP) epitope tag found in several E. coli expression vectors.
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 801
Test Specificity Recombinant proteins containing the calmodulin binding protein epitope tag.
Target Species Other
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.