Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Antibody (3H4A7), Novus Biologicals™
SDP

Catalog No. NBP147546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147546 0.1 mL
NBP147546SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP147546 Supplier Novus Biologicals Supplier No. NBP147546
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

Calnexin Monoclonal specifically detects Calnexin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Calnexin
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3H4A7
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Formulation Ascites with 0.03% Sodium Azide
Gene Accession No. P27824
Gene Alias calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Gene Symbols CANX
Host Species Mouse
Immunogen A synthetic peptide (CEAAEERPWLWVVYILTVAL) corresponding to a region of human Calnexin, conjugated to KLH. [UniProt# P27824]
Molecular Weight of Antigen 90 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ER Markers, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 821
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.