Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Antibody (AF18), HRP, Novus Biologicals™
SDP

Catalog No. NB300518H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300518H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300518H Supplier Novus Biologicals Supplier No. NB300518H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calnexin Monoclonal specifically detects Calnexin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Calnexin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AF18
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Alias calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Gene Symbols CANX
Host Species Mouse
Immunogen Focus human hepatoma cell lysate.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
Primary or Secondary Primary
Gene ID (Entrez) 821
Test Specificity Calnexin (AF18)
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.