Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N101965A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N101965A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N101965A594 Supplier Novus Biologicals Supplier No. NB1001965AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calnexin Polyclonal specifically detects Calnexin in Avian, Bovine, Chicken, Drosophila, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus species, Zebrafish samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Calnexin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Gene Symbols CANX
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the canine Calnexin protein (within residues 25-100). [Swiss-Prot P24643]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 821
Target Species Avian, Bovine, Chicken, Drosophila, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus species, Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.