Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB1001974R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001974R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001974R Supplier Novus Biologicals Supplier No. NB1001974R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calnexin Polyclonal specifically detects Calnexin in Hamster, Human, Mouse, Rat, Zebrafish samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Calnexin
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Gene Symbols CANX
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the rat Calnexin protein (between residues 550-591) [Uniprot: P35565]
Molecular Weight of Antigen 97 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ER Markers, Neuroscience, Sensory Systems, Unfolded Protein Response, Vision
Primary or Secondary Primary
Gene ID (Entrez) 821
Test Specificity Rat, human, and mouse. Hamster reactivity reported in the scientific literature (PMID: 23760268). Predicted to react with Xenopus laevis, dog, orangutan, cow, chicken, Drosophilia melanogaster, guinea pig, quail, pig, rabbit, and sheep based on 100% sequence homology.
Target Species Hamster, Human, Rat, Mouse, Rat, Zebrafish
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.