Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Antibody (IE2.1C12), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NBP236571J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236571J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP236571J Supplier Novus Biologicals Supplier No. NBP236571APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calnexin Monoclonal specifically detects Calnexin in Human samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen Calnexin
Applications Flow Cytometry
Classification Monoclonal
Clone IE2.1C12
Conjugate APC
Dilution Flow Cytometry, Flow (Intracellular)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P27824
Gene Alias calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Gene Symbols CANX
Host Species Mouse
Immunogen Partial recombinant human Calnexin protein (between amino acids 1-300) [UniProt# P27824]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
Primary or Secondary Primary
Gene ID (Entrez) 821
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.