Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Antibody (IE2.1C12) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP236571 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236571 0.1 mg
NBP236571SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP236571 Supplier Novus Biologicals Supplier No. NBP236571
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Calnexin Monoclonal specifically detects Calnexin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen Calnexin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone IE2.1C12
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry 7 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 7 ug/ml, Flow (Intracellular) 5 ug/million cells
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P27824
Gene Alias calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Gene Symbols CANX
Host Species Mouse
Immunogen Partial recombinant human Calnexin protein (between amino acids 1-300) [UniProt# P27824]
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
Primary or Secondary Primary
Gene ID (Entrez) 821
Test Specificity Immunogen's sequence similarity with other species: Mouse (88%), Duck (81%), Xenopus (76%)
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.