Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Antibody, Novus Biologicals™
SDP

Catalog No. NB018779 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018779 0.025 mg
NBP19747605 0.05 mg
NBP19747602 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018779 Supplier Novus Biologicals Supplier No. NBP1974760.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Calnexin Polyclonal specifically detects Calnexin in Human, Mouse, Rat, Porcine, Bovine, Canine, C. elegans, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Single Cell Western.

Specifications

Antigen Calnexin
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Alias calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Gene Symbols CANX
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence near the N-terminus of canine calnexin.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 821
Target Species Rabbit, Sheep, Bovine, Canine, Hamster, Pig, Primate, Rat, Xenopus species, Mouse, C. elegans, Human, Guinea Pig, Chicken
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.