Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 15 Antibody, Novus Biologicals™
SDP

Catalog No. NBP232129 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232129 Supplier Novus Biologicals Supplier No. NBP232129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calpain 15 Polyclonal specifically detects Calpain 15 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen Calpain 15
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 10 mcg/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_005623.1
Gene Alias calpain-15, CAPN15Small optic lobes homolog, EC 3.4.22, EC 3.4.22.-, EC 3.4.22.54, MGC131491, small optic lobes (Drosophila) homolog, small optic lobes homolog (Drosophila)
Gene Symbols SOLH
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 350 to 400 of human Small Optic Lobes Homolog using the numbering given in entry NP_005623.1 (GeneID 6650).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 6650
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.