Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 5 Antibody (OTI7D2), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289675 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289675 Supplier Novus Biologicals Supplier No. NBP270328APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calpain 5 Monoclonal antibody specifically detects Calpain 5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Calpain 5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI7D2
Conjugate APC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias calpain 5, Calpain htra-3, calpain-5, EC 3.4.22, EC 3.4.22.54, FLJ46245, HTRA3NCL3, nCL-3EC 3.4.22.-, New calpain 3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 274-578 of human CAPN5 (NP_004046) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 726
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.