Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain S1 Antibody (1D11), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB161505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB161505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB161505 Supplier Novus Biologicals Supplier No. NBP259446IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calpain S1 Monoclonal antibody specifically detects Calpain S1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Calpain S1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1D11
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Calcium-activated neutral proteinase small subunit, Calcium-dependent protease small subunit 1, calcium-dependent protease, small subunit, calpain 4, small subunit (30K), Calpain regulatory subunit, calpain small subunit 1, calpain, small polypeptide, CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1, epididymis secretory sperm binding protein
Host Species Mouse
Immunogen Recombinant human Calpain S1 (84-268aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 826
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.